Assay Details
| Gene Symbol: C4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002805.2 | NP_001002805.1 | 25 - 26 | 3250 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
inflammatory response
complement activation
biological_process
negative regulation of endopeptidase activity
immunoglobulin mediated immune response
detection of molecule of bacterial origin
positive regulation of apoptotic cell clearance
complement binding
complement component C1q binding
molecular_function
endopeptidase inhibitor activity
carbohydrate binding
inflammatory response
complement activation
biological_process
negative regulation of endopeptidase activity
immunoglobulin mediated immune response
detection of molecule of bacterial origin
positive regulation of apoptotic cell clearance
complement binding
complement component C1q binding
molecular_function
endopeptidase inhibitor activity
carbohydrate binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 3833 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 40356 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10828264 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828256 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828261 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828263 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828250 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828252 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828268 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828266 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828273 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828253 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828276 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828260 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828258 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828269 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828254 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828275 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828259 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828255 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828265 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828251 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828257 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828274 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828271 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828267 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828270 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828262 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828272 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10828229 | - | - |
| Rat Genome 230 2.0 Array | 1370892_at | - | - |
| Rat Genome 230 2.0 Array | 1370892_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR2000014793.rn.1 | TC2000001815.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000014800.rn.1 | TC2000001815.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000014814.rn.1 | TC2000001815.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000014818.rn.1 | TC2000001815.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000014794.rn.1 | TC2000001815.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000014811.rn.1 | TC2000001815.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000014820.rn.1 | TC2000001815.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000014821.rn.1 | TC2000001815.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000014809.rn.1 | TC2000001815.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000014812.rn.1 | TC2000001815.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000014819.rn.1 | TC2000001815.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000014815.rn.1 | TC2000001815.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000014798.rn.1 | TC2000001815.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000014805.rn.1 | TC2000001815.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000014806.rn.1 | TC2000001815.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000014807.rn.1 | TC2000001815.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000014808.rn.1 | TC2000001815.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000014813.rn.1 | TC2000001815.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000014797.rn.1 | TC2000001815.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000014801.rn.1 | TC2000001815.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000014816.rn.1 | TC2000001815.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000014817.rn.1 | TC2000001815.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000014796.rn.1 | TC2000001815.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000014803.rn.1 | TC2000001815.rn.1 | RTA TC |
| Rat Expression Set 230 | 1370892_PM_at | - | - |
| Rat Expression Set 230 | 1370892_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10828229 | - | - |
| Rat Genome U34 Set | U42719_at | - | - |
| Rat Genome U34 Set | rc_AA800942_at | - | - |
| Rat Genome U34 Set | U42719_at | - | - |
| Rat Genome U34 Set | rc_AA800942_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20976862 | - | - |
| ABI Rat Genome Survey Microarray | 21016929 | - | - |
| ABI Rat Genome Survey Microarray | 21622987 | - | - |
| ABI Rat Genome Survey Microarray | 22022582 | - | - |
| ABI Rat Genome Survey Microarray | 22112951 | - | - |
| ABI Rat Genome Survey Microarray | 22124190 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361837 | - | - |

C4b
Genomic Map