Assay Details
| Gene Symbol: G4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003975.3 | NP_001003975.1 | 3 | 886 | 93 | |
| XM_006256073.2 | XP_006256135.1 | 2 | 1097 | 93 | ||
| GenBank mRNA | BC101859.1 | - | 3 | 866 | 7937508 | 93 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

G4
Genomic Map