Assay Details
| Gene Symbol: G6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003976.1 | NP_001003976.1 | 1 - 2 | 68 | 77 | |
| XM_006256069.2 | XP_006256131.1 | 1 - 2 | 620 | 77 | ||
| XM_008772746.1 | XP_008770968.1 | 1 - 2 | 620 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

G6b
Genomic Map