Assay Details
| Gene Symbol: Apom | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019373.2 | NP_062246.1 | 2 - 3 | 348 | 81 | |
| GenBank mRNA | AF207821.1 | - | 2 - 3 | 325 | 81 | |
| BC089807.1 | - | 2 - 3 | 297 | 7323885 | 81 | |
| FQ210093.1 | - | 2 - 3 | 347 | 81 | ||
| FQ219449.1 | - | 2 - 3 | 348 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Apom
Genomic Map