Assay Details
| Gene Symbol: Fmr1nb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006229540.2 | XP_006229602.1 | 3 - 4 | 618 | 135 | |
| XM_008773613.1 | XP_008771835.1 | 4 - 5 | 636 | 135 | ||
| XM_215239.7 | XP_215239.2 | 4 - 5 | 636 | 135 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Fmr1nb
Genomic Map