Assay Details
| Gene Symbol: Snx9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127637.1 | NP_001121109.1 | 13 - 14 | 1797 | 60 | |
| GenBank mRNA | BC158878.1 | - | 13 - 14 | 1797 | 7935618 | 60 |
More Information
Set Membership: |
|

Snx9
Genomic Map