Assay Details
| Gene Symbol: Tmem181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130939.1 | NP_001124411.1 | 2 - 3 | 209 | 55 | |
| GenBank mRNA | BC168231.1 | - | 2 - 3 | 209 | 7387910 | 55 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P047201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559259 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371704 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840951 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196688 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101382 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856651 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712706 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841420 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134191 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499181 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19521 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102498 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158045 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707842 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154079 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32039 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169804 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872906 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882205 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122100 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98655 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P047201 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P047201 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P047201 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P10650 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10702890 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702889 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702896 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702885 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702884 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702894 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702888 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702886 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702883 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702887 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702895 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702881 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702891 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702893 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702882 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10702880 | - | - |
| Rat Genome 230 2.0 Array | 1384428_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1384428_at | - | - |
| Affymetrix HT Rat Focus Array | 1384428_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1384428_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100003011.rn.1 | TC0100000491.rn.1 | RTA TC |
| Rat Expression Set 230 | 1384428_PM_at | - | - |
| Rat Expression Set 230 | 1384428_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702880 | - | - |
| Rat Genome U34 Set | rc_AA955305_at | - | - |
| Rat Genome U34 Set | rc_AI235681_f_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22078122 | - | - |

Tmem181
Genomic Map