Assay Details
| Gene Symbol: Tagap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003748695.3 | XP_003748743.1 | 4 - 5 | 632 | 70 | |
| XM_006227912.2 | XP_006227974.1 | 3 - 4 | 158 | 70 | ||
More Information
Set Membership: |
|

Tagap
Genomic Map