Assay Details
| Gene Symbol: Tulp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109137.1 | NP_001102607.1 | 7 - 8 | 1257 | 67 | |
| XM_006227887.2 | XP_006227949.1 | 8 - 9 | 3610 | 67 | ||
| XM_008758736.1 | XP_008756958.1 | 8 - 9 | 3094 | 67 | ||
| XM_008758737.1 | XP_008756959.1 | 8 - 9 | 3782 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tulp4
Genomic Map