Assay Details
| Gene Symbol: Fzd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044251.2 | NP_001037716.1 | 1 | 2720 | 71 | |
| XM_006254142.2 | XP_006254204.1 | 2 | 4148 | 71 | ||
| GenBank mRNA | BC100088.1 | - | 1 | 2723 | 7371338 | 71 |
| Gene Symbol: LOC100909849 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751721.3 | XP_003751769.1 | 4148 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fzd8
Genomic Map