Assay Details
| Gene Symbol: Pdss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751734.3 | XP_003751782.2 | 10 - 11 | 2206 | 60 | |
| XM_008771923.1 | XP_008770145.1 | 10 - 11 | 2246 | 60 | ||
| GenBank mRNA | BC158695.1 | - | 10 - 11 | 1086 | 60 | |
| FQ220359.1 | - | 7 - 8 | 676 | 60 | ||
More Information
Set Membership: |
|

Pdss1
Genomic Map