Assay Details
| Gene Symbol: Potec | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006254371.2 | XP_006254433.2 | 2 - 3 | 1533 | 98 | |
| XM_008771919.1 | XP_008770141.1 | 1533 | 98 | |||
| XM_008771920.1 | XP_008770142.1 | 1533 | 98 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Potec
Genomic Map