Assay Details
| Gene Symbol: Krit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108618.1 | NP_001102088.1 | 10 - 11 | 1591 | 76 | |
| GenBank mRNA | BC168923.1 | - | 10 - 11 | 1598 | 7105532 | 76 |
More Information
Updated Mapping Information
Set Membership: |
|

Krit1
Genomic Map