Assay Details
Gene Symbol: Lrrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_216085.7 | XP_216085.3 | 4 - 5 | 2943 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
