Assay Details
| Gene Symbol: Atp5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023093.1 | NP_075581.1 | 9 - 10 | 1331 | 104 | |
| GenBank mRNA | BC061830.1 | - | 9 - 10 | 1331 | 5599635 | 104 |
| FQ214146.1 | - | 8 - 9 | 1173 | 104 | ||
| FQ225416.1 | - | 4 - 5 | 646 | 104 | ||
| FQ225735.1 | - | 4 - 5 | 646 | 104 | ||
| FQ227531.1 | - | 4 - 5 | 646 | 104 | ||
| FQ227720.1 | - | 3 - 4 | 466 | 104 | ||
| J05266.1 | - | 9 - 10 | 1252 | 104 | ||
| X56133.1 | - | 2 - 3 | 328 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp5a1
Genomic Map