Assay Details
| Gene Symbol: Rpl41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139083.2 | NP_620783.1 | 1 - 2 | 70 | 62 | |
| GenBank mRNA | FQ217332.1 | - | 1 - 2 | 70 | 62 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the NM_139083.2 transcripts.
This assay is in the 5'UTR region of the NM_139083.2 transcripts.
Set Membership: |
|

Rpl41
Genomic Map