Assay Details
| Gene Symbol: Ormdl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105940.1 | NP_001099410.1 | 3 - 4 | 539 | 77 | |
| XM_006240753.2 | XP_006240815.1 | 3 - 4 | 519 | 77 | ||
| XM_006240754.2 | XP_006240816.1 | 3 - 4 | 439 | 77 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ormdl2
Genomic Map