Assay Details
| Gene Symbol: Clpp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_217313.8 | XP_217313.6 | 4 - 5 | 2259 | 75 | |
| GenBank mRNA | FQ231323.1 | - | 4 - 5 | 546 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Clpp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_217313.8 | XP_217313.6 | 4 - 5 | 2259 | 75 | |
| GenBank mRNA | FQ231323.1 | - | 4 - 5 | 546 | 75 | |
Set Membership: |
|