Assay Details
| Gene Symbol: LOC100909732 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003752157.3 | XP_003752205.1 | 1053 | 85 | ||
| XM_006257660.2 | XP_006257722.1 | 1053 | 85 | |||
| XM_006257665.2 | XP_006257727.1 | 1053 | 85 | |||
| XM_008773657.1 | XP_008771879.1 | 1053 | 85 | |||
| XM_008773658.1 | XP_008771880.1 | 1053 | 85 | |||
| Gene Symbol: Fam122b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166586.1 | NP_001160058.1 | 2 - 3 | 513 | 85 | |
| XM_006257643.2 | XP_006257705.1 | 2 - 3 | 1053 | 85 | ||
| XM_006257648.2 | XP_006257710.1 | 2 - 3 | 1053 | 85 | ||
| XM_008773633.1 | XP_008771855.1 | 2 - 3 | 1053 | 85 | ||
| XM_008773634.1 | XP_008771856.1 | 2 - 3 | 1053 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

LOC100909732
Genomic Map