Assay Details
| Gene Symbol: Hprt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012583.2 | NP_036715.1 | 1 - 2 | 94 | 165 | |
| GenBank mRNA | BC098629.1 | - | 1 - 2 | 94 | 7386548 | 165 |
| M63983.1 | - | 1 - 2 | 126 | 165 | ||
| S79292.1 | - | 1 - 2 | 126 | 165 | ||
| X62085.1 | - | 54 | 165 | |||
| Gene Symbol: LOC103689983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773659.1 | XP_008771881.1 | 1 - 2 | 170 | 165 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC098629.1,M63983.1,NM_012583.2,S79292.1,X62085.1,XM_008773659.1 transcripts.
This assay is in the 5'UTR region of the BC098629.1,M63983.1,NM_012583.2,S79292.1,X62085.1,XM_008773659.1 transcripts.
Set Membership: |
|

Hprt1
Genomic Map