Assay Details
| Gene Symbol: Npepo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012346.1 | NP_001012346.1 | 1 - 2 | 794 | 100 | |
| XM_008771429.1 | XP_008769651.1 | 3 - 4 | 1242 | 100 | ||
| XM_008771430.1 | XP_008769652.1 | 3 - 4 | 1242 | 100 | ||
| XM_008771431.1 | XP_008769653.1 | 2 - 3 | 1089 | 100 | ||
| XM_008771432.1 | XP_008769654.1 | 2 - 3 | 1081 | 100 | ||
| XM_008771433.1 | XP_008769655.1 | 2 - 3 | 954 | 100 | ||
| GenBank mRNA | AJ810421.1 | - | 1 - 2 | 794 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Npepo
Genomic Map