Assay Details
| Gene Symbol: Npepo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012346.1 | NP_001012346.1 | 8 - 9 | 1872 | 88 | |
| XM_008771429.1 | XP_008769651.1 | 10 - 11 | 2326 | 88 | ||
| XM_008771430.1 | XP_008769652.1 | 10 - 11 | 2326 | 88 | ||
| XM_008771431.1 | XP_008769653.1 | 9 - 10 | 2173 | 88 | ||
| XM_008771432.1 | XP_008769654.1 | 9 - 10 | 2165 | 88 | ||
| XM_008771433.1 | XP_008769655.1 | 9 - 10 | 2038 | 88 | ||
| GenBank mRNA | AJ810421.1 | - | 8 - 9 | 1872 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Npepo
Genomic Map