Assay Details
| Gene Symbol: Npepo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012346.1 | NP_001012346.1 | 14 - 15 | 2308 | 107 | |
| XM_008771429.1 | XP_008769651.1 | 16 - 17 | 2774 | 107 | ||
| XM_008771430.1 | XP_008769652.1 | 16 - 17 | 2774 | 107 | ||
| XM_008771431.1 | XP_008769653.1 | 15 - 16 | 2621 | 107 | ||
| XM_008771432.1 | XP_008769654.1 | 15 - 16 | 2613 | 107 | ||
| XM_008771433.1 | XP_008769655.1 | 15 - 16 | 2486 | 107 | ||
| GenBank mRNA | AJ810421.1 | - | 14 - 15 | 2308 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P231867 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950144 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872959 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103346 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846479 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366084 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283545 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823590 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628702 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789671 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75595 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377924 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707265 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195155 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889346 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423366 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344552 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929102 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500036 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534185 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663699 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542885 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789681 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430935 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250177 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41227 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823697 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288281 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335976 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334479 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613625 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307307 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455577 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327039 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945760 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816166 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166799 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714565 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461154 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753277 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531510 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384420 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474521 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581957 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388362 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465294 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432311 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654820 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108535 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219181 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562236 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858121 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314831 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55362 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283197 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 995 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877328 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813346 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769914 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193745 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880688 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106751 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939407 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267714 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796932 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16919 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29951 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152463 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127123 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118666 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158992 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78625 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130975 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8354 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127191 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89992 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 180223 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143675 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158484 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113512 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P231867 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P231867 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P231867 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P231867 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P431354 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36939 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 98731 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 85546 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 45436 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 11056 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 92835 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 70892 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 3274 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 22957 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36874 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 95996 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 31287 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 80788 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 50439 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 3568 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 67073 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 91912 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 53658 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 27300 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 30941 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 23309 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 103239 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10796992 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797003 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797004 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797001 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796995 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797000 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796994 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797005 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796999 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797006 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797002 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796997 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796996 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796993 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796998 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10796991 | - | - |
| Rat Genome 230 2.0 Array | 1374767_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1374767_at | - | - |
| Affymetrix HT Rat Focus Array | 1374767_at | - | - |
| Affymetrix HT Rat Focus Array | 1374767_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1700000082.rn.1 | TC1700000013.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374767_PM_at | - | - |
| Rat Expression Set 230 | 1374767_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796991 | - | - |
| Rat Genome U34 Set | rc_AA799396_g_at | - | - |
| Rat Genome U34 Set | rc_AA799396_at | - | - |
| Rat Genome U34 Set | rc_AI072900_at | - | - |
| Rat Genome U34 Set | AA799397_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21634828 | - | - |
| ABI Rat Genome Survey Microarray | 21826465 | - | - |

Npepo
Genomic Map