Assay Details
| Gene Symbol: Npepo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008771429.1 | XP_008769651.1 | 13 - 14 | 2498 | 62 | |
| XM_008771430.1 | XP_008769652.1 | 13 - 14 | 2498 | 62 | ||
| XM_008771431.1 | XP_008769653.1 | 12 - 13 | 2345 | 62 | ||
| XM_008771432.1 | XP_008769654.1 | 12 - 13 | 2337 | 62 | ||
| XM_008771433.1 | XP_008769655.1 | 12 - 13 | 2210 | 62 | ||
More Information
Set Membership: |
|

Npepo
Genomic Map