Assay Details
| Gene Symbol: Zkscan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025760.1 | NP_001020931.1 | 1 - 2 | 112 | 105 | |
| XM_006249015.2 | XP_006249077.1 | 1 - 2 | 155 | 105 | ||
| GenBank mRNA | BC099191.1 | - | 1 - 2 | 112 | 7383463 | 105 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC099191.1, NM_001025760.1, XM_006249015.2 transcripts.
Set Membership: |
|

Zkscan1
Genomic Map