Assay Details
| Gene Symbol: Zkscan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025760.1 | NP_001020931.1 | 2 - 3 | 609 | 64 | |
| XM_006249015.2 | XP_006249077.1 | 2 - 3 | 652 | 64 | ||
| GenBank mRNA | BC099191.1 | - | 2 - 3 | 609 | 7383463 | 64 |
More Information
Updated Mapping Information
Set Membership: |
|

Zkscan1
Genomic Map