Assay Details
Gene Symbol: RGD1305455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024969.2 | NP_001020140.2 | 7 - 8 | 1234 | 81 | |
GenBank mRNA | CO394313.1 | - | 2 - 3 | 179 | 81 | |
FQ211718.1 | - | 7 - 8 | 1280 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
