Assay Details
Gene Symbol: RGD1305455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024969.2 | NP_001020140.2 | 9 - 10 | 1551 | 85 | |
GenBank mRNA | BC091263.1 | - | 7 - 8 | 1257 | 85 | |
CO394313.1 | - | 4 - 5 | 496 | 85 | ||
FQ211718.1 | - | 9 - 10 | 1597 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
