Assay Details
| Gene Symbol: RGD1305455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024969.2 | NP_001020140.2 | 5 - 6 | 1004 | 74 | |
| GenBank mRNA | BC091263.1 | - | 4 - 5 | 874 | 74 | |
| DY317491.1 | - | 372 | 74 | |||
| FQ211718.1 | - | 5 - 6 | 1050 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

RGD1305455
Genomic Map