Assay Details
| Gene Symbol: Taf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044225.1 | NP_001037690.1 | 10 - 11 | 1178 | 86 | |
| XM_008769061.1 | XP_008767283.1 | 10 - 11 | 1218 | 86 | ||
| XM_008769062.1 | XP_008767284.1 | 10 - 11 | 1217 | 86 | ||
| GenBank mRNA | BC100108.1 | - | 10 - 11 | 1178 | 7379862 | 86 |
| D49446.1 | - | 9 - 10 | 1149 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Taf6
Genomic Map