Assay Details
| Gene Symbol: Wash | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127390.1 | NP_001120862.1 | 235 | 74 | ||
| GenBank mRNA | BC166761.1 | - | 235 | 7930951 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wash
Genomic Map