Assay Details
| Gene Symbol: Zkscan3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012053.2 | NP_001012053.2 | 1 - 2 | 31 | 75 | |
| GenBank mRNA | BC089847.1 | - | 1 - 2 | 31 | 7373658 | 75 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC089847.1, NM_001012053.2 transcripts.
Set Membership: |
|

Zkscan3
Genomic Map