Assay Details
| Gene Symbol: Aasdhppt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106798.2 | NP_001100268.2 | 4 - 5 | 711 | 174 | |
| GenBank mRNA | BC098729.1 | - | 3 - 4 | 1067 | 174 | |
| BC166800.1 | - | 4 - 5 | 711 | 7938259 | 174 | |
| FQ209549.1 | - | 4 - 5 | 727 | 174 | ||
| FQ214028.1 | - | 4 - 5 | 730 | 174 | ||
| FQ214274.1 | - | 4 - 5 | 724 | 174 | ||
| FQ230080.1 | - | 4 - 5 | 730 | 174 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Aasdhppt
Genomic Map