Assay Details
Gene Symbol: Zfp414 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009664.1 | NP_001009664.1 | 1 - 2 | 111 | 92 | |
XM_006241119.2 | XP_006241181.1 | 1 - 2 | 264 | 92 | ||
GenBank mRNA | BC087693.1 | - | 1 - 2 | 111 | 7310348 | 92 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC087693.1,NM_001009664.1,XM_006241119.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
