Assay Details
Gene Symbol: Zfp414 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009664.1 | NP_001009664.1 | 2 - 3 | 398 | 63 | |
XM_006241117.2 | XP_006241179.1 | 2 - 3 | 588 | 63 | ||
XM_006241119.2 | XP_006241181.1 | 2 - 3 | 551 | 63 | ||
XM_006241121.2 | XP_006241183.1 | 2 - 3 | 586 | 63 | ||
GenBank mRNA | BC087693.1 | - | 2 - 3 | 398 | 7310348 | 63 |
FQ220382.1 | - | 2 - 3 | 442 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
