Assay Details
Gene Symbol: Zfp414 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009664.1 | NP_001009664.1 | 3 - 4 | 503 | 123 | |
XM_006241117.2 | XP_006241179.1 | 3 - 4 | 693 | 123 | ||
XM_006241119.2 | XP_006241181.1 | 3 - 4 | 656 | 123 | ||
XM_006241121.2 | XP_006241183.1 | 3 - 4 | 691 | 123 | ||
GenBank mRNA | BC087693.1 | - | 3 - 4 | 503 | 7310348 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
