Assay Details
| Gene Symbol: Cass4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191744.1 | NP_001178673.1 | 1 - 2 | 37 | 83 | |
| XM_006235679.2 | XP_006235741.1 | 1 - 2 | 940 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cass4
Genomic Map