Assay Details
Gene Symbol: Rab22a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108966.1 | NP_001102436.1 | 5 - 6 | 702 | 118 | |
XM_006235705.2 | XP_006235767.1 | 6 - 7 | 878 | 118 | ||
GenBank mRNA | BC158650.1 | - | 6 - 7 | 702 | 8367507 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P713239 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P122110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658175 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650144 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689415 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607845 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734369 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828593 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732874 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422448 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502332 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746490 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346821 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152494 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828357 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968107 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691389 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971979 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759289 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192118 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771795 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99318 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3037 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111297 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P713239 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P122110 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P713239 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P122110 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P122110 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P713239 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P310787 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P121325 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 90778 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 99585 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10842615 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10842619 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10842618 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10842617 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10842620 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10842616 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10842614 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10842613 | - | - |
Rat Genome 230 2.0 Array | 1372862_at | - | - |
Rat Genome 230 2.0 Array | 1389692_at | - | - |
Rat Genome 230 2.0 Array | 1377896_at | - | - |
Rat Genome 230 2.0 Array | 1389692_PM_at | - | - |
Rat Genome 230 2.0 Array | 1372862_PM_at | - | - |
Rat Genome 230 2.0 Array | 1377896_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1372862_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1377896_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1377896_at | - | - |
Affymetrix HT Rat Focus Array | 1372862_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300018476.rn.1 | TC0300002504.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300018475.rn.1 | TC0300002504.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300018473.rn.1 | TC0300002504.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300018477.rn.1 | TC0300002504.rn.1 | RTA TC |
Rat Expression Set 230 | 1372862_at | - | - |
Rat Expression Set 230 | 1372862_PM_at | - | - |
Rat Expression Set 230 | 1389692_at | - | - |
Rat Expression Set 230 | 1389692_PM_at | - | - |
Rat Expression Set 230 | 1377896_at | - | - |
Rat Expression Set 230 | 1377896_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10842613 | - | - |
Rat Genome U34 Set | rc_AI030866_at | - | - |
Rat Genome U34 Set | rc_AA818769_at | - | - |
Rat Genome U34 Set | rc_AI172383_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20700370 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353287 | - | - |
