Assay Details
| Gene Symbol: Pmepa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107807.2 | NP_001101277.1 | 1 - 2 | 117 | 84 | |
| XM_006235689.1 | XP_006235751.1 | 2 - 3 | 282 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pmepa1
Genomic Map