Assay Details
| Gene Symbol: Ctsz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183330.1 | NP_899159.1 | 2 - 3 | 395 | 101 | |
| GenBank mRNA | AB023781.2 | - | 2 - 3 | 395 | 101 | |
| BC091110.1 | - | 2 - 3 | 329 | 7379777 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ctsz
Genomic Map