Assay Details
| Gene Symbol: Abcb1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006235994.2 | XP_006236056.1 | 17 - 18 | 2775 | 106 | |
| GenBank mRNA | AY582535.1 | - | 14 - 15 | 1865 | 106 | |
More Information
Set Membership: |
|

Abcb1a
Genomic Map