Assay Details
| Gene Symbol: Abcb1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133401.1 | NP_596892.1 | 4 - 5 | 340 | 87 | |
| XM_006235994.2 | XP_006236056.1 | 6 - 7 | 1165 | 87 | ||
| GenBank mRNA | AF257746.1 | - | 4 - 5 | 340 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abcb1a
Genomic Map