Assay Details
| Gene Symbol: Suox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031127.3 | NP_112389.3 | 1 - 2 | 70 | 133 | |
| XM_006240802.2 | XP_006240864.1 | 2 - 3 | 150 | 133 | ||
| XM_006240803.2 | XP_006240865.1 | 3 - 4 | 251 | 133 | ||
| GenBank mRNA | BC061991.1 | - | 1 - 2 | 70 | 5599452 | 133 |
More Information
Updated Mapping Information
Set Membership: |
|

Suox
Genomic Map