Assay Details
| Gene Symbol: Pa2g4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004206.1 | NP_001004206.1 | 4 - 5 | 513 | 98 | |
| GenBank mRNA | BC079095.1 | - | 4 - 5 | 513 | 7124036 | 98 |
More Information
Updated Mapping Information
Set Membership: |
|

Pa2g4
Genomic Map