Assay Details
| Gene Symbol: Pa2g4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004206.1 | NP_001004206.1 | 6 - 7 | 666 | 92 | |
| GenBank mRNA | BC079095.1 | - | 6 - 7 | 666 | 7124036 | 92 |
| FQ233061.1 | - | 6 - 7 | 674 | 92 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pa2g4
Genomic Map