Assay Details
| Gene Symbol: Pa2g4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004206.1 | NP_001004206.1 | 8 - 9 | 824 | 74 | |
| GenBank mRNA | BC079095.1 | - | 8 - 9 | 824 | 7124036 | 74 |
| FQ233061.1 | - | 8 - 9 | 832 | 74 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pa2g4
Genomic Map