Assay Details
| Gene Symbol: Ptges3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130989.1 | NP_001124461.1 | 1 - 2 | 295 | 158 | |
| XM_008765026.1 | XP_008763248.1 | 1 - 2 | 332 | 158 | ||
| GenBank mRNA | BC166579.1 | - | 1 - 2 | 295 | 7379926 | 158 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC166579.1, NM_001130989.1, XM_008765026.1 transcripts.
Set Membership: |
|

Ptges3
Genomic Map