Assay Details
| Gene Symbol: G6pd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017006.2 | NP_058702.1 | 3 - 4 | 271 | 93 | |
| GenBank mRNA | BC081820.1 | - | 3 - 4 | 184 | 7108556 | 93 |
| X07467.1 | - | 3 - 4 | 197 | 93 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

G6pd
Genomic Map