Assay Details
| Gene Symbol: Dusp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037973.1 | NP_001033062.1 | 1 - 2 | 186 | 126 | |
| GenBank mRNA | BC110044.1 | - | 1 - 2 | 186 | 7462125 | 126 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC110044.1, NM_001037973.1 transcripts.
Set Membership: |
|

Dusp9
Genomic Map