Assay Details
| Gene Symbol: Emd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012948.1 | NP_037080.1 | 3 - 4 | 381 | 143 | |
| GenBank mRNA | BC059151.1 | - | 3 - 4 | 394 | 6919042 | 143 |
| X98377.1 | - | 3 - 4 | 381 | 143 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Emd
Genomic Map